Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMIGO3 Monoclonal Antibody (371301), Invitrogen™
Rat Monoclonal Antibody
Supplier: Thermo Scientific MA524114
Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse AMIGO2, recombinant human (rh) AMIGO1, or rhAMIGO2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
AMIGO3 Monoclonal antibody specifically detects AMIGO3 in Mouse samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Specifications
AMIGO3 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8C2S7 | |
Amigo3 | |
Mouse myeloma cell line NS0-derived recombinant mouse AMIGO3 Thr20-Pro378 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Immunohistochemistry (Frozen), Western Blot | |
371301 | |
Unconjugated | |
Amigo3 | |
adhesion molecule with Ig like domain 3; adhesion molecule with Ig-like domain 3; ALI3; alivin 3; alivin-3; AMIGO3; AMIGO-3; amigo3 {ECO:0000312; amphoterin induced gene and ORF 3; amphoterin-induced gene and ORF 3; amphoterin-induced protein 3; E430002N15Rik; EMBL:AAO48952.1, ECO:0000312; Kiaa1851; MGC120552; mKIAA1851; RGD:631413}; transmembrane protein AMIGO3; UNQ6084/PRO20089 | |
Rat | |
Protein A/G | |
RUO | |
320844 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction