Learn More
anti-Aminopeptidase N/CD13, Alexa Fluor 700, Clone: WM-15, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB8284N
Catalog No. FAB8284N
Add to cart
This item is not returnable. View return policy
Description
Description
Aminopeptidase N/CD13 Monoclonal antibody specifically detects Aminopeptidase N/CD13 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Aminopeptidase N/CD13
WM-15
Flow Cytometry 5 uL/10^6 cells
Purified
ANPEP
Human AML cells
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
290
Human
Flow Cytometry
Alexa Fluor 700
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
alanyl (membrane) aminopeptidase, Alanyl aminopeptidase, Aminopeptidase M, aminopeptidase N, ANPEP, AP-M, APN, AP-N, CD13, CD13 antigen, CD13APN, EC 3.4.11, EC 3.4.11.2, gp150, LAP1, Microsomal aminopeptidase, Myeloid plasma membrane glycoprotein CD13, P150, PEPN, PEPNhAPN
Mouse
IgG1
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human Aminopeptidase N/CD13 in flow cytometry