Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-AMOT, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215378
Description
AMOT Polyclonal specifically detects AMOT in Human samples. It is validated for Western Blot.Specifications
AMOT | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
angiomotin, angiomotin p80 isoform, KIAA1071angiomotin p130 isoform | |
Rabbit | |
118 kDa | |
0.1 mL | |
Primary | |
Human. Expected to cross react based on sequence identity: Bovine (98%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
Q4VCS5 | |
AMOT | |
Recombinant protein encompassing a sequence within the center region of human AMOT. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
154796 | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only