Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Amylase 2B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524187

Catalog No. PIPA524187


Add to cart

Description

Description

Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch andglycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas. This gene encodes an amylase isoenzyme produced by the pancreas.
Specifications

Specifications

alpha Amylase 2B
Polyclonal
PBS with 0.09% sodium azide
P19961
AMY2, 1,4-alpha-D-glucan glucanohydrolase 2B, alpha-amylase 2B, alpha-amylase carcinoid, carcinoid alpha-amylase, glycogenase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
280
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
AMY2B
Liquid
AMY2B
KLH conjugated synthetic peptide between 74-100 amino acids from the N-terminal region of human AMY2B
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit