Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyloid A Mouse anti-Human, Clone: Reu-86.2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA135013

Catalog No. 01-677-055


Add to cart

Description

Description

The serum amyloid A (SAA) family comprises a number of differentially expressed lipoproteins, acute-phase SAA1 and SAA2, the former being a major component in plasma, and constitutive SAA ft.s (C-SAAs). Although the liver is the primary site of synthesis of both SAA types, extrhepatic production has been reported. The in-vivo concentrations increase by as much as 1000 fold during inflammation. Several studies have expressed it ft.s importance in the diagnosis and monitoring of various diseases.
Specifications

Specifications

Amyloid A
Monoclonal
Unconjugated
SAA1
Liquid
SAA1
Human serum Amyloid-A.
1 mL
4° C
Primary
6288
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Reu-86.2
tissue culture supernatant with 0.1% sodium azide
P0DJI8
REU-86.2
Mouse
IgG2a
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

This product is for in vitro experimental use only