Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyloid Β (AΒ)x-40 Mouse anti-Human, Mouse, Clone: 11A5-B10, MilliporeSigma™

Mouse Monoclonal Antibody

Manufacturer:  MilliporeSigma 05799

Catalog No. 05-799-MI


Add to cart

Description

Description

Detect Amyloid β (Aβ)x-40 using this Anti-Amyloid β (Aβ)x-40 Antibody, clone 11A5-B10 validated for use in western blot, immunohistochemistry.
Specifications

Specifications

Amyloid β (Aβ)x-40
11A5-B10
mouse ascites containing 0.05% sodium azide
Ascites
Murine
IgG
100μL
Primary
Human, Murine
Immunohistochemistry, Western Blot
Unconjugated
NM_000484
APP, ABETA, CTFgamma, ABPP, PreA4, PN-II, AD1, APPI, A4, CVAP, PN2, AAA
peptide corresponding to amino acids 34-40 of human Amyloid βx-40 peptide
Ascites
−20°C for 24 months.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only