Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Amyloid Fibrils OC, Polyclonal, ™ Non-distribution product as customer accommodation.

Manufacturer:  MilliporeSigma™ AB2286/DEL

Catalog No. 50-173-389

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

Amyloid monomeric proteins can sometimes oligomerize into destructive amyloid fibrils. Amyloidogenic conformations of non-disease related proteins can be created by partial protein misfolding or denaturation. In disease state oligomerization, extensive amyloid oligomerization creates plaques in neural tissue that correlates with Alzheimer's symptomology.

Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Human
Immunogen: Fibrils prepared from human Aβ42 peptide.

Dot Blotting, ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Amyloid Fibrils OC
Serum
Fibrils prepared from human Aβ42 peptide.
100μL
Polyclonal
Unconjugated
Rabbit
IgG
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit