Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amyloid Oligomers Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP197930
Description
Amyloid Oligomers Polyclonal antibody specifically detects Amyloid Oligomers in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin).Specifications
Amyloid Oligomers | |
Polyclonal | |
Unconjugated | |
PBS, 50% Glycerol with 0.09% Sodium Azide | |
eukaryotic translation initiation factor 3 subunit p42 | |
Rabbit | |
Protein A purified | |
RUO | |
351 | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.5mg/mL | |
Western Blot 1:1000, ELISA 0.1-10 ug/ml, Immunohistochemistry 1:1000-1:10000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:1000, Immunohistochemistry-Paraffin 1:1000-1:10000 | |
P05067 | |
APP | |
Synthetic molecular mimic of soluble oligomers | |
0.1 mL | |
Primary | |
Recognizes all types of amyloid oligomers. Appears to recognize a peptide backbone epitope that is common to amyloid oligomers, but is not found in native proteins, amyloidogenic monomer or mature amyloid fibrils. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only