Learn More
Amyloid Precursor Protein Rabbit anti-Human, Mouse, Non-human primate, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA133107
Catalog No. PIPA133107
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
PA1-33107 detects APP from human, monkey, and mouse samples. PA1-33107 has been successfully used in immunoprecipitation and Western blot applications.
Amyloid Precursor Protein (APP) or Amyloid beta protein functions as a cell surface kinesin I membrane receptor, mediating the axonal transport of beta-secretase and presenilin 1; APP is rapidly internalized via clathrin-coated pits. APP is important for neurite growth, neuronal adhesion and axonogenesis. Mutations in the APP gene can lead to Alzheimer disease 1 (AD1). This anitbody recognizes APP (b-APP770) with a MW of approximately 100kDa.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Amyloid Precursor Protein
Polyclonal
APP
APP
Proprietary immunogen
RUO
11820, 351
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunoprecipitation, Western Blot
Unconjugated
P05067, P12023
Rabbit
50 μL
Primary
Human, Mouse, Non-human Primate
Antibody
IgG
Recommendations
Recommendations