Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyloid Precursor Protein Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112528

Catalog No. 01-674-188


Add to cart

Description

Description

A suggested positive control for this product is murine brain lysate.

Amyloid Precursor Protein (APP) or Amyloid beta protein functions as a cell surface kinesin I membrane receptor, mediating the axonal transport of beta-secretase and presenilin 1; APP is rapidly internalized via clathrin-coated pits. APP is important for neurite growth, neuronal adhesion and axonogenesis. Mutations in the APP gene can lead to Alzheimer disease 1 (AD1). This anitbody recognizes APP (b-APP770) with a MW of approximately 100kDa.
Specifications

Specifications

Amyloid Precursor Protein
Polyclonal
PBS with 0.02% sodium azide
P05067, P12023, P08592
Amyloid beta A4 protein, ABPP, Alzheimer disease amyloid protein, Cerebral vascular amyloid peptide, CVAP; Protease nexin-II, PN-II, APPI, PreA4, A4,
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
11820, 351, 54226
Western Blot
Unconjugated
APP
Liquid
APP
Synthetic peptide corresponding to residues 737-751 of human amyloid A4 protein precursor.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit