Learn More
anti-ANGEL2, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP130807
Catalog No. NBP130807
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
ANGEL2
LYOPH
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
ANGEL2
A synthetic peptide from the n-terminal region of human ANGEL2 (Protein angel homolog 2) conjugated to an immunogenic carrier protein was used as the immunogen.
IgG purified
RUO
Primary
90806
Reconstitute with 0.5 ml sterilized water.
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Unconjugated
No buffer with No Preservative
angel homolog 2 (Drosophila), KIAA0759LFLJ12793, protein angel homolog 2
Rabbit
IgG
0.5 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Appears to be specific for ANGEL2.
Human