Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-2 Mouse anti-Human, Clone: 85816, R&D Systems™

Mouse Monoclonal Antibody

$114.50 - $839.00

Specifications

Antigen Angiopoietin-2
Immunogen Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-2 Asp68-Phe496 Accession # O15123
Clone 85816
Applications Immunohistochemistry, ELISA
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB098SP
View Documents
R&D Systems
MAB098SP
25 ug
Each for $114.50
Add to cart
 
MAB098100
View Documents
R&D Systems
MAB098100
100 ug
Each for $377.00
Add to cart
 
MAB098500
View Documents
R&D Systems
MAB098500
500 ug
Each for $839.00
Add to cart
 
Description

Description

Angiopoietin-2 Monoclonal antibody MAB098 Specifically detects Angiopoietin-2 in Human samples. It is validated for ELISA Capture
Specifications

Specifications

Angiopoietin-2
85816
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
IgG2a
285
RUO
Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-2 Asp68-Phe496 Accession # O15123
Immunohistochemistry, ELISA
Purified
O15123
Monoclonal
Primary
Detects human and mouse Angiopoietin-2 in ELISAs. In ELISAs, no cross-reactivity or interference with recombinant human (rh) Ang-1, rhAng-4, rhAng-X, recombinant mouse (rm) Ang-3, rmANGPTL-3, rhTie-1, rhTie-2, and rmTie-2 is observed.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit