Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin 2 Goat anti-Human, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA120178

Catalog No. PA120178


Add to cart

Description

Description

PA1-20178 detects Angiopoietin 2 in human samples. PA1-20178 has been successfully used in ELISA and Western blot procedures. The PA1-20178 immunogen is recombit, full length, human Angiopoietin 2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Angiopoietin 2 and angiopoietin 1 have an N-terminal coiled-coil domain and a C-terminal fibrinogen-like domain. The coiled coil domains mediate ligand homo-oligomerization and the fibrinogen-like domains mediate ligand activity. Recombit human angiopoietin 2 has a molecular mass of approximately 66 kDa in SDS-PAGE under reducing and non-reducing conditions. Angiopoietin 2 probably acts as a natural antagonist for Ang1 and Tie2 both in vivo and in vitro. In adult mice and humans, angiopoietin 2 is expressed at sites of vascular remodeling.
Specifications

Specifications

Angiopoietin 2
Polyclonal
Unconjugated
ANGPT2
Liquid
ANGPT2
Recombit, full length, human Angiopoietin 2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
0.1 mg/mL
PBS with 5% trehalose and no preservative; pH 7.4
O15123
AGPT2, ANG2
Goat
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
285
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit