Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin 2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527299

Catalog No. PIPA527299


Add to cart

Description

Description

PA5-27299 targets Angiopoietin 2 in IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-27299 immunogen is recombit fragment corresponding to a region within amino acids 1 and 308 of Human Angiopoietin 2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is an antagonist of angiopoietin 1 (ANGPT1) and endothelial TEK tyrosine kinase (TIE-2, TEK). The encoded protein disrupts the vascular remodeling ability of ANGPT1 and may induce endothelial cell apoptosis. Three transcript variants encoding three different isoforms have been found for this gene.
Specifications

Specifications

Angiopoietin 2
Polyclonal
Unconjugated
ANGPT2
Liquid
ANGPT2
Recombit fragment corresponding to a region within amino acids 1 and 308 of Human Angiopoietin 2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1.44 mg/mL
PBS with 20% glycerol and 0.025% Proclin 300; pH 7
O15123
AGPT2, ANG2, ANG-2, Tie2-ligand, angiopoietin-2, angiopoietin-2B, angiopoietin-2a
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
285
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit