Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-like Protein 2/ANGPTL2 Mouse anti-Human, Clone: 239829, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $839.00

Specifications

Antigen Angiopoietin-like Protein 2/ANGPTL2
Immunogen Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-like 2 Ile233-Phe492 Accession # Q9UKU9
Clone 239829
Applications Western Blot
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB2084SP
View Documents
R&D Systems
MAB2084SP
25 ug
Each for $119.60
Add to cart
 
MAB2084100
View Documents
R&D Systems
MAB2084100
100 ug
Each for $377.00
Add to cart
 
MAB2084500
View Documents
R&D Systems
MAB2084500
500 ug
Each for $839.00
Add to cart
 
Description

Description

Angiopoietin-like Protein 2/ANGPTL2 Monoclonal antibody MAB2084 Specifically detects Angiopoietin-like Protein 2/ANGPTL2 in Human samples. It is validated for Western Blotting
Specifications

Specifications

Angiopoietin-like Protein 2/ANGPTL2
239829
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
IgG2b
23452
RUO
Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-like 2 Ile233-Phe492 Accession # Q9UKU9
Western Blot
Purified
Q9UKU9
Monoclonal
Primary
Detects human Angiopoietin-like 2 in direct ELISAs and Western blots. Shows 25-100% cross-reactivity with recombinant mouse Angiopoietin-like 2 and no cross-reactivity with recombinant human (rh) Angiopoietin-1 or rhAngiopoietin-2.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit