Learn More
Angiotensin I Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136039
Catalog No. 01-672-228
Description
Description
Reconstitute with 1 mL of distilled water and add preservative if preferred.
The protein encoded by this gene, pre-angiotensinogen or angiotensinogen precursor, is expressed in the liver and is cleaved by the enzyme renin in response to lowered blood pressure. The resulting product, angiotensin I, is then cleaved by angiotensin converting enzyme (ACE) to generate the physiologically active enzyme angiotensin II. The protein is involved in maintaining blood pressure and in the pathogenesis of essential hypertension and preeclampsia. Mutations in this gene are associated with susceptibility to essential hypertension, and can cause renal tubular dysgenesis, a severe disorder of renal tubular development. Defects in this gene have also been associated with non-familial structural atrial fibrillation, and inflammatory bowel disease.
Specifications
Specifications
Angiotensin I
Polyclonal
whole serum with no preservative
P01019
RABBIT anti Human ANGIOTENSIN I
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
ELISA
Unconjugated
AGT
Lyophilized
AGT
Polyclonal Rabbit anti Angiotensin I antibody.
1 mL
4° C
Primary
183