Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKFY1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524640

Catalog No. PIPA524640


Add to cart

Description

Description

ANKFY1 encodes a cytoplasmic protein that contains a coiled-coil structure and a BTB/POZ domain at its N-terminus, ankyrin repeats in the middle portion, and a FYVE-finger motif at its C-terminus. This protein belongs to a subgroup of double zinc finger proteins which may be involved in vesicle or protein transport.
Specifications

Specifications

ANKFY1
Polyclonal
PBS with 0.09% sodium azide
Q9P2R3
ANKHZN, ZFYVE14, Rabankyrin-5, ankyrin repeat and FYVE domain-containing protein 1, ankyrin repeat hooked to zinc finger motif, ankyrin repeats hooked to a zinc finger motif
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51479
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ANKFY1
Liquid
ANKFY1
KLH conjugated synthetic peptide between 436-465 amino acids from the C-terminal region of human ANKFY1
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit