Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ANKHD1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178211

Catalog No. NBP178211


Add to cart

Description

Description

ANKHD1 Polyclonal antibody specifically detects ANKHD1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

ANKHD1
1.0 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 mcg/mg
NP_060217.1
ankyrin repeat and KH domain containing 1, FLJ10042, FLJ11979, FLJ14127, hMASK, KIAA1085single KH-domain homolog, MASKFLJ20288, Multiple ankyrin repeats single KH domain
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
ANKHD1
The immunogen for this product maps to a region between residue 25 and 75 of human Ankyrin Repeat and KH Domain Containing 1 using the numbering given in entry NP_060217.1 (GeneID 54882).
Immunogen affinity purified
RUO
Primary
54882
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only