Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD32 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525322

Catalog No. PIPA525322


Add to cart

Specifications

Specifications

ANKRD32
Polyclonal
PBS with 0.09% sodium azide
Q9BQI6, Q8R3P9
BRCTD1, BRCTx, BRCT domain containing 1, BRCT domain-containing protein 1, ankyrin repeat domain-containing protein 32
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
105377, 84250
Immunohistochemistry, Western Blot
Unconjugated
SLF1
Liquid
SLF1
KLH conjugated synthetic peptide between 159-188 amino acids from the N-terminal region of human ANKRD32
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit