Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD57 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525701

Catalog No. PIPA525701

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ANKRD57
Polyclonal
PBS with 0.09% sodium azide
Q53LP3
ANKRD57, C2orf26, ankyrin repeat domain 57, ankyrin repeat domain-containing protein 57
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
65124
Immunohistochemistry, Western Blot
Unconjugated
SOWAHC
Liquid
SOWAHC
KLH conjugated synthetic peptide between 479-507 amino acids from the C-terminal region of human ANKRD57
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit