Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA137083
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is placenta tissue.
Annexin A1, a gene related to phagocytosis, was found to be one of the most highly upregulated genes in hairy cell leukemia.Specifications
Annexin A1 | |
Polyclonal | |
ANXA1 | |
ANX1, LPC1, annexin I lipocortin I, annexin-1, calpactin II, calpactin-2, chromobindin-9, lipocortin I, p35, phospholipase A2 inhibitory protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
301 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
P04083 | |
ANXA1 | |
Synthetic peptide corresponding to N-terminus of human annexin 1 | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction