Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin II Light Chain Mouse, Unlabeled, Clone: 148, BD

Mouse Monoclonal Antibody

$207.00 - $419.00

Specifications

Antigen Annexin II Light Chain
Isotype IgG1
Immunogen Cow Annexin II Light Chain aa. 1-97
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610070
View Documents
BD Biosciences
610070
50μg Each for $207.00
Add to cart
 
BDB610071
View Documents
BD Biosciences
610071
150μg Each for $419.00
Add to cart
 
Description

Description

Annexin II often exists as a heterotetramer complexed with an 11kDa light chain. This p11 subunit is related to the S100 family of calcium-binding proteins but mutations have rendered it unable to bind calcium. The p11 protein binds to the amino terminus of the annexin II monomer, but not to other annexins. It may regulate the function of annexin II, as the tetrameric form seems to have a higher calcium and phospholipid-binding affinity than the monomeric annexin II. In the cell, p11 has been localized to the cytoplasmic face of the plasma membrane where the annexin II tetramer is known to exist.

Host Species: Mouse
Clone: 148
Isotype: IgG1
Species Reactivity: Human
Immunogen: Cow Annexin II Light Chain aa. 1-97
Formula Weight [Chemical]: 11kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Annexin II Light Chain
Cow Annexin II Light Chain aa. 1-97
Unconjugated
Cell Biology
Murine
Affinity Purified
Monoclonal
Store undiluted at -20°C.
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
148
250μg/mL

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit