Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin VI Mouse, Unlabeled, Clone: 73, BD

Mouse Monoclonal Antibody

$220.00 - $405.00

Specifications

Antigen Annexin VI
Isotype IgG1
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610300
View Documents
BD Biosciences
610300
50μg Each for $220.00
Add to cart
 
BDB610301
View Documents
BD Biosciences
610301
150μg Each for $405.00
Add to cart
 
Description

Description

Annexins are a widely expressed family of Ca2+ and phospholipid-binding proteins. At least ten have been identified in mammalian tissues. They have also been identified in Drosophila, Hydra, and Dictyostelium. Annexin VI is a 70kDa member of the annexin family. While most of the other annexin family members have a four amino acid sequence repeat, Annexin VI displays an eight residue repeat. It is a widely expressed protein, found in lymphocytes, neurons, and many other cell types. Work on annexins has addressed the possible role of Annexin VI in various stages of the endocytic pathway. Some studies suggest that Annexin VI is required for coated-vesicle budding. However, other research directly contradicts this data. Thus, the role of Annexin VI is still under investigation.

Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

Annexin VI
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Unconjugated
Cell Biology
Murine
Affinity Purified
Monoclonal
Store undiluted at -20°C.
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
73
250μg/mL

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit