Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin A6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137086

Catalog No. 01-673-062


Add to cart

Description

Description

This antibody is predicted to react with bovine, canine, mouse, porcine based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is liver tissue.

The Annexin family of calcium-binding proteins is composed of al least ten mammalian genes. It is characterized by a conserved core domain which binds to phospholipids in a Ca2+-dependent manner and a unique amino terminal region which may confer binding specificity. The Annexin family has been implicated as regulators of such diverse processes as ion-flux, endocytosis and exocytosis, and cellular adhesion. When overexpressed in A431 cells, Annexin VI causes a partial reversal of the transformed phenotype.
Specifications

Specifications

Annexin A6
Polyclonal
PBS with 1% BSA and <0.1% sodium azide; pH 7.4
P08133
ANX6, CBP68, 67 kDa calelectrin, CPB-II, annexin VI p68, annexin-6, calcium-binding protein p68, calelectrin, calphobindin II, calphobindin-II, chromobindin-20, lipocortin VI, p68, p70
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
309
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ANXA6
Liquid
ANXA6
Synthetic peptide corresponding to N-terminus of human Annexin VI.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit