Learn More
KAL1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA521149
Catalog No. PIPA521149
Add to cart
Description
Description
A suggested positive control is MCF7 cell lysate. PA5-21149 can be used with blocking peptide PEP-1263.
Mutations in Anosmin-1, an extracellular matrix-associated glycosylated protein, have been linked with Kallmann Syndrome (KS), an X-linked genetic disorder characterized by loss of smell caused by abnormal olfactory bulb development and delayed puberty caused by disrupted migration of the gonadotropin-releasing hormone neuron. Anosmin-1 has been shown to directly bind FGFR1 via its N-terminal cysteine-rich domain, whey-acidic protein-like domain, and its first FnIII repeat with the D2 and D3 ectodomains of FGFR1. It is thought that Anosmin-1 can modulate FGFR1 signaling and have opposing effects on the formation and activation of FGF2-FGFR1-heparing complex.
Specifications
Specifications
KAL1
Polyclonal
Unconjugated
ANOS1
Liquid
ANOS1
An 18 amino acid peptide near the center of human Anosmin.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P23352
ADMLX, HHA, KAL, KALIG-1, KMS; Kallmann syndrome interval gene 1; Kallmann syndrome-1 sequence (anosmin-1); adhesion molecule-like X-linked; anosmin-1; kallmann syndrome protein
Rabbit
IgG
100 μg
4° C
Primary
3730