Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANR44 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524266

Catalog No. PIPA524266

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Putative regulatory subunit of protein phospatase 6 (PP6) that may be involved in the recognition of phosphoprotein substrates.
Specifications

Specifications

ANR44
Polyclonal
PBS with 0.09% sodium azide
Q8N8A2
PP6-ARS-B, ankyrin repeat domain-containing protein 44, protein phosphatase 6 ankyrin repeat subunit B, serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit B, serine/threonine-protein phosphatase 6 regulatory subunit ARS-B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
91526
Immunohistochemistry, Western Blot
Unconjugated
ANKRD44
Liquid
ANKRD44
KLH conjugated synthetic peptide between 481-510 amino acids from the central region of human ANR44
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit