Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANR52 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524295

Catalog No. PIPA524295

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with non-human primate based on sequence homology.

Putative regulatory subunit of protein phospatase 6 (PP6) that may be involved in the recognition of phosphoprotein substrates.
Specifications

Specifications

ANR52
Polyclonal
PBS with 0.09% sodium azide
Q8NB46
ANKRD33, CVWG5837, PP6-ARS-C, ankyrin repeat domain 33, ankyrin repeat domain-containing protein 52, protein phosphatase 6 ankyrin repeat subunit C, serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit C, serine/threonine-protein phosphatase 6 regula
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
283373
Flow Cytometry, Western Blot
Unconjugated
ANKRD52
Liquid
ANKRD52
KLH conjugated synthetic peptide between 433-461 amino acids from the N-terminal region of human ANR52
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit