Learn More
Anthrax Protective Antigen Rabbit anti-Bacteria, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA141695
Catalog No. 01-675-100
Add to cart
Description
Description
Anthrax PA (Protective Antigen) and Anthrax LF (Lethal Factor), along with Anthrax EF (Edema Factor), are toxins produced by Bacillus anthracis. All three of the proteins must be present for toxic entry into cells.  PA is secreted from Bacillus anthracis as an 83 kDa (PA83) protein which is cleaved by furin protease, releasing a 20 kDa fragment. The 63 kDa portion  (PA63) of the cleavage product forms a heptamer, which binds the toxins LF and EF. This complex enters via receptor-mediated endocytosis where LF and EF exert their toxic effects.
Specifications
Specifications
Anthrax Protective Antigen
Polyclonal
whole serum with no preservative
Anthrax PA
Synthetic peptides corresponding to residues 186-200 of the Anthrax Pa.
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
Bacteria
Western Blot
Unconjugated
Liquid
Rabbit
IgG
RUO
Antibody
Polyclonal