Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Annexin A3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA512438
Description
ANXA3 is a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions in the inhibition of phopholipase A2 and cleavage of inositol 1,2-cyclic phosphate to form inositol 1-phosphate. This protein may also play a role in anti-coagulation.Specifications
Annexin A3 | |
Polyclonal | |
ANXA3 | |
Annexin A3; Annexin-3; Annexin III; Lipocortin III; Placental anticoagulant protein III; PAP-III; 35-alpha calcimedin; Inositol 1,2-cyclic phosphate 2-phosphohydrolase; ANXA3; ANX3 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
306 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
P12429 | |
ANXA3 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human ANXA3 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title