Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin A3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512439

Catalog No. PIPA512439

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ANXA3 is a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions in the inhibition of phopholipase A2 and cleavage of inositol 1,2-cyclic phosphate to form inositol 1-phosphate. This protein may also play a role in anti-coagulation.
Specifications

Specifications

Annexin A3
Polyclonal
PBS with 0.09% sodium azide
P12429, O35639
Annexin A3; Annexin-3; Annexin III; Lipocortin III; Placental anticoagulant protein III; PAP-III; 35-alpha calcimedin; Inositol 1,2-cyclic phosphate 2-phosphohydrolase; ANXA3; ANX3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11745, 306
Western Blot
Unconjugated
ANXA3
Liquid
ANXA3
KLH conjugated synthetic peptide between 146-178 amino acids from the central region of human ANXA3
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit