Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Annexin A7, Polyclonal, Invitrogen™

Catalog No. PIPA127778
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127778 50 μg
1 options

Catalog No. PIPA127778

Supplier: Thermo Scientific PA127778

Chicken Polyclonal Antibody

Calcium/phospholipid-binding protein which promotes membrane fusion and is involved in exocytosis. Annexin VII is a member of the annexin family of calcium-dependent phospholipid binding proteins. The Annexin VII gene contains 14 exons and spans approxima

PA1-27778 detects ANXA7 from human samples.PA1-27778 may be successfully used in Western blot applications.The PA1-27682 immunogen is a synthetic peptide corresponding to human Annexin A7.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANXA7
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene ANXA7
Gene Accession No. P20073
Gene Alias RP11-537A6.8, ANX7, SNX, SYNEXIN
Gene Symbols ANXA7
Host Species Chicken
Immunogen Synthetic peptide corresponding to human Annexin A7.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.