Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AOC3, Mouse, Clone: 4B8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant AOC3.
Supplier: Abnova Corporation H00008639M06
Description
Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor. The product is a major protein on the adipocyte plasma membrane. It has adhesive properties and also has functional monoamine oxidase activity. A pseudogene for this gene has been discribed and is located approximately 9kb downstream. [provided by RefSeq
Sequence: DVRFQGERLVYEISLQEALAIYGGNSPAAMTTRYVDGGFGMGKYTTPLTRGVDCPYLATYVDWHFLLESQAPKTIRDAFCVFEQNQGLPLRRHHSDLYSHSpecifications
AOC3, | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_003734 | |
AOC3 | |
AOC3 (NP_003725, 351 a.a. ∼ 450 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunofluorescence | |
4B8 | |
Mouse monoclonal antibody raised against a partial recombinant AOC3. | |
AOC3 | |
HPAO/SSAO/VAP-1/VAP1 | |
Mouse | |
Affinity chromatography | |
RUO | |
8639 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction