Learn More
MilliporeSigma™ anti-AP-2γ, Clone: 6E4/4,
Manufacturer:  MilliporeSigma™ 05909
Catalog No. 05-909-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
AP-2γ
6E4/4
NM_003222.3
TFAP2C, AP2-GAMMA, TFAP2G, hAP-2g, AP2-gamma, ERF1
Bacterially expressed AP-2 protein
Protein G purfied
−20°C for up to 24 months.
Monoclonal
Immunohistochemistry, Western Blot
Unconjugated
Purified
Murine
IgG
100μg
Primary
Human, Murine