Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma Adaptin Goat anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131775

Catalog No. PA131775


Add to cart

Description

Description

PA1-31775 detects AP1G1 from human samples. PA1-31775 has been successfully used in Western blot applications. The PA1-31775 immunogen is synthetic peptide: CTAPTSKPSSAGGEL, corresponding to amino acids 646 - 659 of Human AP1G1.

Adaptins are important components of clathrin-coated vesicles transporting ligand-receptor complexes from the plasma membrane or from the trans-Golgi network to lysosomes. The adaptin family of proteins is composed of four classes of molecules named alpha, beta-, beta prime- and gamma- adaptins. Adaptins, together with medium and small subunits, form a heterotetrameric complex called an adaptor, whose role is to promote the formation of clathrin-coated pits and vesicles. The protein encoded by this gene is a gamma-adaptin protein and it belongs to the adaptor complexes large subunits family. Two transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

gamma Adaptin
Polyclonal
Unconjugated
AP1G1
Liquid
AP1G1
synthetic peptide CTAPTSKPSSAGGEL, corresponding to amino acids 646 - 659 of Human AP1G1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
0.7 mg/mL
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
O43747, P22892
ADTG, CLAPG1, MGC18255
Goat
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
11765, 164, 171494
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit