Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP2M1, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Supplier:  Thermo Scientific PA127945

Catalog No. PIPA127945

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

PA1-27945 detects AP2M1 from human and mouse samples.PA1-27945 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27922 immunogen is a synthetic peptide corresponding to human AP-2 complex subunit mu-1.Store at 4ºC short term. For extended storage, aliquot and store at -20ºC for short term. Avoid freeze/thaw cycles.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AP2M1
Polyclonal
Unconjugated
Q96CW1, P84091
AP2M1
Synthetic peptide corresponding to human AP-2 complex subunit mu-1.
50 μg
Primary
Human, Mouse
Antibody
IgY
Western Blot
1mg/mL
AP2M1
AP50, CLAPM1, mu2
Chicken
Antigen affinity chromatography
RUO
1173
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Purified
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit