Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP3S2, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127781

Catalog No. PIPA127781

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Part of the AP-3 complex. an adapter-related complex which is not clathrin-associated. The complex is associated with the Golgi region as well as more peripheral structures. It facilitates the budding of vesicles from the Golgi membrane and may be directl

PA1-27781 detects AP3S2 from human and mouse samples.PA1-27781 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27758 immunogen is a synthetic peptide corresponding to human AP-3 complex subunit sigma-2.
Specifications

Specifications

AP3S2
Polyclonal
PBS with no preservative
P59780, Q8BSZ2
AP3S3, FLJ35955, sigma3b
Chicken
IgY
50μg
Store at -20°C
Primary
10239
Western Blot
Unconjugated
AP3S2
Purified
AP3S2
Synthetic peptide corresponding to human AP-3 complex subunit sigma-2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit