Learn More
anti-APAF-1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP176999
Catalog No. NBP176999
Add to cart
Description
Description
APAF-1 Polyclonal antibody specifically detects APAF-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
APAF-1
1 mg/mL
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
AAC51678
APAF-1apoptotic protease activating factor 1, apoptotic peptidase activating factor, apoptotic peptidase activating factor 1, apoptotic protease activating factor, apoptotic protease-activating factor 1, CED4, DKFZp781B1145, KIAA0413
Rabbit
IgG
Peptide affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PBS with 0.02% Sodium Azide
Affinity Purified
APAF1
Antibody was raised against a peptide corresponding to amino acids near the carboxy terminus of human Apaf1. The sequence of the immunogenic peptide differs from that of murine Apaf1 by one amino acid. The immunogen is located within the last 50 amino acids of Apaf1.
130 kDa
0.1 mg
Apoptosis, Cancer, Cell Biology, Mitochondrial Markers, Tumor Suppressors
Primary
317