Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein APC, Polyclonal, Invitrogen™

Catalog No. PIPA121004
Change view
Click to view available options
Quantity:
500 μL
Catalog No. Quantity
PIPA121004 500 μL
1 options

Catalog No. PIPA121004

Supplier: Thermo Scientific PA121004

Rabbit Polyclonal Antibody

This gene encodes a tumor suppressor protein that includes among its many intracellular functions one of nuclear export. Defects in this gene cause familial adenomatous polyposis (FAP), an autosomal dominant pre-malignant disease that usually progresses to malignancy. Disease-associated mutations tend to be clustered in a small region designated the mutation cluster region (MCR) and result in a truncated protein product.

PA1-21004 detects APC in Human and Mouse samples.PA1-21004 has been successfully used in Immunohistochemistry (Paraffin) procedures.PA1-21004 immunogen corresponds to Synthetic peptide corresponding to the C-terminal region of the human protein. .
TRUSTED_SUSTAINABILITY

Specifications

Antigen APC
Applications Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.2mg/mL
Conjugate Unconjugated
Gene APC
Gene Accession No. P25054, Q61315
Gene Alias Adenomatous Polyposis Coli, APC, DP2, DP2.5, DP3, FAP, FPC, GS
Gene Symbols APC
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal region of the human protein.
Purification Method Antigen affinity chromatography
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 324
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C, do not freeze
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.