Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APIP Mouse anti-Human, Clone: 19F424, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA141380

Catalog No. 01-676-618


Add to cart

Description

Description

Suggested positive control: antigen standard for APIP (transient overexpression lysate), recombit protein.

The mammalian homologues of the key cell death gene CED-4 in C. elegans has been identified recently from human and mouse and designated Apaf1 (for apoptosis protease-activating factor 1) (1,2). Apaf1 binds to cytochrome c (Apaf2) and caspase-9 (Apaf3), which leads to caspase-9 activation. Activated caspase-9 in turn cleaves and activates caspase-3 that is one of the key proteases, being responsible for the proteolytic cleavage of many key proteins in apoptosis3. Recently, Cho et al (4) have identified a new Apaf-1 Interactiong Protein (APIP) also known as CG129 and MMRP19, as a negative regulator of ischemic injury. APIP competes with Caspase-9 binding site of Apaf1. APIP is predicted to code for a 204 amino acid. An isoform of APIP, APIP2 encodes a 242 amino acid protein, which is an alternative splicing variant differing in its N-terminus from APIP. APIP transcript is ubiquitously expressed in most adult tissue with high expression in skeletal muscle, heart, and kidney.
Specifications

Specifications

APIP
Monoclonal
0.5 mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
Q96GX9
CG129, MMRP19, APIP
Mouse
IgG1, kappa
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
51074
Western Blot
19F424
Unconjugated
APIP
Liquid
APIP
Full-length His-tagged recombit APIP protein.
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit