Learn More
ATG10 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA118292
Catalog No. 01-670-545
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-18292 detects APG10 from human samples. PA1-18292 has been successfully used in immunohistochemistry and immunofluorescence procedures. The PA1-18292 immunogen is a synthetic peptide corresponding to the C-terminal residues of human APG10 protein. Reconstitute with 100 μL of distilled water.
Autophagy is a process for the bulk degradation of cytosolic compartments by lysosomes. ATG10 is an E2-like enzyme involved in 2 ubiquitin-like modifications essential for autophagosome formation: ATG12 .-ATG5 . conjugation and modification of a soluble form of MAP-LC3 (MAP1LC3A; MIM 601242), a homolog of yeast Apg8, to a membrane-bound form (Nemoto et al., 2003 [PubMed 12890687]).
Specifications
Specifications
ATG10
Polyclonal
whole serum with no preservative
Q9H0Y0
ATG10
Synthetic peptide corresponding to the C-terminal residues of human APG10 protein.
RUO
83734
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
Immunofluorescence, Immunohistochemistry
Unconjugated
ATG10
Autophagy-related protein 10, APG10-like, ATG10
Rabbit
100 μL
Primary
Human
Antibody
IgG