Learn More
ATG12 Rabbit anti-Human, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA118284
Catalog No. 01-670-537
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-18284 detects APG12 from human and rat samples. PA1-18284 has been successfully used in immunohistochemistry, immunofluorescence, and Western blot procedures. The PA1-18284 immunogen is a synthetic peptide corresponding to the N-terminal residues of human APG12 protein Reconstitute with 500 μL of distilled water.
APG12 is required for autophagy. APG5 conjugates to ATG12 and associates with isolation membrane to form cup-shaped isolation membrane and autophagosome. The conjugate detaches from the membrane immediately before or after autophagosome formation is completed.
Specifications
Specifications
ATG12
Polyclonal
Unconjugated
ATG12
Lyophilized
ATG12
Synthetic peptide corresponding to the N-terminal residues of human APG12 protein
Protein G
RUO
Antibody
Polyclonal
Human, Rat
Immunofluorescence, Immunohistochemistry, Western Blot
1 mg/mL
Antibody with no preservative
O94817, Q2TBJ5
Autophagy-related protein 12, APG12-like, APG 12, ATG12, APG12, APG12L
Rabbit
IgG
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
361321, 9140