Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG16L1 Rabbit anti-Human, Mouse, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523217

Catalog No. PIPA523217


Add to cart

Description

Description

Autophagy is a process for the bulk degradation of cytosolic compartments by lysosomes/vacuoles. The process is found in eukaryotes, including yeast, and involves several genes and two novel ubiquitin-like conjugation systems. The APG12 system is one of the ubiquitin-like protein conjugation systems essential for autophagy. Covalent attachment of APG12–APG5 is essential for autophagy. In mammalian cells, the APG12–APG5 conjugate is a protein complex of approximately 800 kDa and includes an additional novel protein. This novel protein was designated APG16L (APG16-like, also referred to as ATG16L) as it ft.s N-terminal region contains several features similar to yeast APG16. APG16L protein has several spliced isoforms and is larger than yeast APG16 (17 kD). The protein has a large C-terminal domain containing seven WD repeats. Genetic variations in the protein are associated with susceptibility to inflammatory bowel disease type 10. The human APG16L gene is localized in the chromosomal region 2q37.1.
Specifications

Specifications

ATG16L1
Polyclonal
Unconjugated
ATG16L1
Liquid
ATG16L1
Synthetic peptide (55 KLQAEKHDVPNRHEISPG 72) of APG16L protein.
Protein G
RUO
Antibody
Polyclonal
Human, Mouse, Non-human primate
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q676U5, Q8C0J2
hCG_1817841, APG16L, ATG16A, ATG16L, IBD10, WDR30, APG16L beta, WD repeat domain 30, autophagy-related protein 16-1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
55054, 77040
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit