Learn More
ATG4B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSA00033W
Catalog No. PIOSA00033W
Add to cart
Description
Description
Reconstitute with 150 µL of distilled water.
QKI belongs to a family of RNA-binding proteins that have an HNRNPK. QKI proteins regulate RNA splicing, export of target RNAs from the nucleus, translation of proteins, and RNA stability.
Specifications
Specifications
ATG4B
Polyclonal
whole serum with no preservative
Q9Y4P1, Q8BGE6
Cysteine protease ATG4B, autophagin-1, AUTL1
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry, Western Blot
Unconjugated
ATG4B
Lyophilized
ATG4B
A synthetic peptide from aa region 20-80 of human ATG4B conjugated to an immunogenic carrier protein has been used as the antigen
150 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
23192, 66615