Learn More
ApoA-1 Mouse anti-Rat, Unlabeled, Clone: 4, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 612331
 View more versions of this product
Catalog No. BDB612331
Add to cart
Description
Description
In blood, cholesterol and triglycerides are transported in lipoprotein particles that consist of a single layer of phospholipid surrounding a lipid core and surface-associated apolipoproteins (Apo). The Apo-proteins are involved in the specific binding of cellular receptors, the regulation of lipolytic enzymes, and the process of lipid exchange. High density lipoprotein particles (HDLs) contain the apolipoproteins ApoA-I, ApoA-II and Apo-M, whereas low density (LDL), intermediate density, and very low density (VLDL) lipoprotein particles contain ApoB-100 as the primary structural element. ApoA-I is synthesized in the liver and small intestines, where it acts as a cofactor for lecithin-cholesterol acyltransferase during the formation of cholesterol ester. ApoA-II is synthesized in the liver where it activates hepatic lipase. In HepG2 cells, treatment with gramoxone causes oxidative stress and reductions in ApoA-I mRNA levels. This down regulation of ApoA-I may contribute to reduced plasma HDL levels in response to oxidants, such as cigarette smoke. Thus, ApoA-I is an important cofactor fo cholesterol synthesis and is a major component of HDLs.
Western Blotting
Specifications
Specifications
ApoA-1
4
Unconjugated
Murine
IgG1
RUO
Primary
Rat
Western Blot
250μg/mL
Affinity Purified
Rat ApoA-I aa. 144-258
150μg
Cell Biology
Monoclonal
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only.