Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein A5 Rabbit anti-Chicken, Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521168

Catalog No. PIPA521168


Add to cart

Description

Description

A suggested positive control is human liver tissue lysate. PA5-21168 can be used with blocking peptide PEP-1282.

Apolipoprotein A4 (also known as ApoA-IV) is a plasma protein that is O-linked glycoprotein after proteolytic processing. ApoA4 is a major constituent of HDL and it also can be found on the surface of newly synthesized chylomicrons. The physiological function of ApoA4 is not completely identified so far, but several evidences from vitro studies indicate that this apolipoprotein involves in several steps of the reverse cholesterol transport pathway, which transports cholesterol from peripheral cells back to the liver.
Specifications

Specifications

Apolipoprotein A5
Polyclonal
Unconjugated
APOA5
Liquid
APOA5
A 15 amino acid peptide near the carboxy terminus of chicken ApoA5.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Chicken, Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q6Q788
UNQ411/PRO773, APOAV, RAP3; apo-AV; apolipoprotein A5; regeneration-associated protein 3
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
116519, 419802
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit