Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOBEC3B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511430

Catalog No. PIPA511430


Add to cart

Description

Description

PHO3 is a member of the cytidine deaminase gene family. The PHO3 gene is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control.
Specifications

Specifications

APOBEC3B
Polyclonal
PBS with 0.09% sodium azide
Q9UH17, Q99J72
APOBEC3, APOBEC3B
KLH conjugated synthetic peptide between 299-330 amino acids from the C-terminal region of human APOBEC3B (PHO3)
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
Unconjugated
APOBEC3B
Liquid
Rabbit
Ig
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80287, 9582
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit