Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOBEC3G Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511428

Catalog No. PIPA511428

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CEM15 is a member of the cytidine deaminase family. It is the product of one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control. CEM15 has been found to be a specific inhibitor of human immunodeficiency virus-1 (HIV-1) infectivity.
Specifications

Specifications

APOBEC3G
Polyclonal
PBS with 0.09% sodium azide
Q9HC16
DNA dC->dU-editing enzyme APOBEC-3G, APOBEC-related cytidine deaminase, ARCD, APOBEC-related protein, ARP-9, CEM15, CEM-15, APOBEC3G, MDS019
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
60489
Immunohistochemistry, Western Blot
Unconjugated
APOBEC3G
Liquid
APOBEC3G
KLH conjugated synthetic peptide between 352-384 amino acids from the C-terminal region of human APOBEC3G (CEM15)
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit