Learn More
APOC2, Mouse, Clone: 3E4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant APOC2.
Supplier: Abnova Corporation H00000344M01
Description
The protein encoded by this gene is secreted in plasma where it is a component of very low density lipoprotein. This protein activates the enzyme lipoprotein lipase, which hydrolyzes triglycerides and thus provides free fatty acids for cells. Mutations in this gene cause hyperlipoproteinemia type IB, characterized by hypertriglyceridemia, xanthomas, and increased risk of pancreatitis and early atherosclerosis. [provided by RefSeq
Sequence: TQQPQQDEMPSPTLLTQVKESLSSYWESAKTAAQNLYEKTYLPAVDEKLRDLYSKSTAAMSTYTGIFTDQVLSVLKGEESpecifications
APOC2 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC005348 | |
APOC2 | |
APOC2 (AAH05348, 23 a.a. ∼ 101 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
3E4 | |
Mouse monoclonal antibody raised against a partial recombinant APOC2. | |
APOC2 | |
MGC75082 | |
Mouse | |
Affinity chromatography | |
RUO | |
344 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.