Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA514322
Description
This antibody is predicted to react with rat and rabbit based on sequence homology.
Apolipoprotein D Polyclonal antibody specifically detects Apolipoprotein D in Human, Mouse samples. It is validated for Western Blot.Specifications
Apolipoprotein D | |
Polyclonal | |
Unconjugated | |
APOD | |
AOPDGN; APOD; Apo-D; APO-D protein; Apolipoprotein D | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
11815, 347 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P05090, P51910 | |
APOD | |
KLH conjugated synthetic peptide between 26-53 amino acids from the N-terminal region of human APOD | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction