Learn More
APOE Goat anti-Human, Polyclonal Antibody, Abnova
Goat polyclonal antibody raised against recombinant APOE.
Manufacturer: Abnova Corporation PAB14301
Description
Chylomicron remnants and very low density lipoprotein (VLDL) remnants are rapidly removed from the circulation by receptor-mediated endocytosis in the liver. Apolipoprotein E, a main apoprotein of the chylomicron, binds to a specific receptor on liver cells and peripheral cells. ApoE is essential for the normal catabolism of triglyceride-rich lipoprotein constituents. The APOE gene is mapped to chromosome 19 in a cluster with APOC1 and APOC2. Defects in apolipoprotein E result in familial dysbetalipoproteinemia, or type III hyperlipoproteinemia (HLP III), in which increased plasma cholesterol and triglycerides are the consequence of impaired clearance of chylomicron and VLDL remnants. [provided by RefSeq
Specifications
APOE | |
Unconjugated | |
Western Blot (1:5000-1:40000) ELISA (1:5000-1:40000) The optimal working dilution should be determined by the end user. | |
APOE | |
AD2|LPG|MGC1571|apoprotein | |
Goat | |
100 ug | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
This antibody specifically binds to human APOE. |
ELISA, Western Blot | |
Goat polyclonal antibody raised against recombinant APOE. | |
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
Liquid | |
APOE | |
Recombinant protein corresponding to APOE. | |
RUO | |
Primary | |
348 | |
Human |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only